missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-2,4-difluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Supplier:  TCI America C32261G

 View more versions of this product

Catalog No. C32261G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Cyano-2,4-difluorophenylboronic Acid (contains varying amounts of Anhydride)
134°C
C7H4BF2NO2
1 g
VQEKFJVJHSVFTM-UHFFFAOYSA-N
(3-cyano-2,4-difluorophenyl)boronic acid
44558118
Crystalline Powder
871940-31-7
White
MFCD12026699
3-Cyano-2,4-difluorobenzeneboronic Acid
B(C1=C(C(=C(C=C1)F)C#N)F)(O)O
182.921
182.92

Chemical Identifiers

871940-31-7
182.921
VQEKFJVJHSVFTM-UHFFFAOYSA-N
44558118
B(C1=C(C(=C(C=C1)F)C#N)F)(O)O
C7H4BF2NO2
MFCD12026699
3-Cyano-2,4-difluorobenzeneboronic Acid
(3-cyano-2,4-difluorophenyl)boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions