missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-1-ethyl-6-hydroxy-4-methyl-2-pyridone 98.0+%, TCI America™

Catalog No. C278525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C278525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C278525G Supplier TCI America Supplier No. C278525G
Only null left

Chemical Identifiers

CAS 28141-13-1
Molecular Formula C9H10N2O2
Molecular Weight (g/mol) 178.19
MDL Number MFCD00142624
InChI Key YSNMMQRIPFUHAO-UHFFFAOYSA-N
Synonym 3-Cyano-1-ethyl-6-hydroxy-3-picoline, 1-Ethyl-1,2-dihydro-6-hydroxy-4-methyl-2-oxo-3-pyridinecarbonitrile
PubChem CID 119879
IUPAC Name 1-ethyl-2-hydroxy-4-methyl-6-oxo-1,6-dihydropyridine-3-carbonitrile
SMILES CCN1C(=O)C=C(C)C(C#N)=C1O

Specifications

Melting Point 233°C
Color Red-White
Quantity 25 g
UN Number 3439
Formula Weight 178.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyano-1-ethyl-6-hydroxy-4-methyl-2-pyridone
EINECSNumber (5)-3716
TSCA Yes