missing translation for 'onlineSavingsMsg'
Learn More

3-Coumaranone, 97%

Catalog No. p-7080611
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1020203 1 g
AAA1020206 5 g
AAA1020214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1020203 Supplier Thermo Scientific Chemicals Supplier No. A1020203

CAS: 7169-34-8 | C8H6O2 | 134.13 g/mol

3-Coumaranone is used in the aurones, [2-benzylidenebenzofuran-3(2H)-ones] by reacting with appropriate aldehyde. It is involved in the condensation reaction with aldehydes in the presence of morpholine acetate to prepare substituted 2-(arylidene)benzofuran-3(ZH)-ones. In Horner-Wadsworth-Emmons reaction, it reacts with diethyl¬ cyanomethyl¬phosphonate to give 3-(cyanomethyl)benzofurans, which is an intermediate in the synthesis of antihyperglycemic agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Coumaranone is used in the aurones, [2-benzylidenebenzofuran-3(2H)-ones] by reacting with appropriate aldehyde. It is involved in the condensation reaction with aldehydes in the presence of morpholine acetate to prepare substituted 2-(arylidene)benzofuran-3(ZH)-ones. In Horner-Wadsworth-Emmons reaction, it reacts with diethyl¬ cyanomethyl¬phosphonate to give 3-(cyanomethyl)benzofurans, which is an intermediate in the synthesis of antihyperglycemic agents.

Solubility
Slightly miscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 7169-34-8
Molecular Formula C8H6O2
Molecular Weight (g/mol) 134.13
MDL Number MFCD00051810
InChI Key MGKPCLNUSDGXGT-UHFFFAOYSA-N
Synonym benzofuran-3 2h-one, 3-coumaranone, 3 2h-benzofuranone, 1-benzofuran-3 2h-one, coumaranone, benzofuran-3-one, coumaran-3-one, 2,3-dihydrobenzo b furan-3-one, 2,3-dihydro-1-benzofuran-3-one, 2h-benzofuran-3-one
PubChem CID 23556
IUPAC Name 1-benzofuran-3-one
SMILES O=C1COC2=CC=CC=C12

Specifications

Melting Point 97°C to 102°C
Quantity 1 g
Beilstein 115296
Sensitivity Light sensitive
Solubility Information Slightly miscible with water.
Formula Weight 134.13
Percent Purity 97%
Chemical Name or Material 3-Coumaranone
EINECSNumber 000-000-0
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only