missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorotoluene 99.0+%, TCI America™

Catalog No. p-7124344
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C029625G 25 g
C0296500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C029625G Supplier TCI America Supplier No. C029625G

Chemical Identifiers

CAS 108-41-8
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000595
InChI Key OSOUNOBYRMOXQQ-UHFFFAOYSA-N
Synonym 3-chlorotoluene, m-chlorotoluene, benzene, 1-chloro-3-methyl, m-tolyl chloride, 3-chloro-1-methylbenzene, 1-methyl-3-chlorobenzene, toluene, m-chloro, meta-chlorotoluene, unii-5k8172k9tt, chlorotoluene 3-
PubChem CID 7931
IUPAC Name 1-chloro-3-methylbenzene
SMILES CC1=CC=CC(Cl)=C1

Specifications

Melting Point -48°C
Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
UN Number 2238
Boiling Point 162°C
Formula Weight 126.58
Percent Purity ≥99.0% (GC)
Chemical Name or Material 3-Chlorotoluene
EINECSNumber (3)-0039
TSCA Yes