missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorotoluene, 97%

Catalog No. AC160451000
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC160451000 100 mL Glass bottle
AC160452500 250 mL Glass bottle
AC160455000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC160451000 Supplier Thermo Scientific Chemicals Supplier No. 160451000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-41-8 | C7H7Cl | 126.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108-41-8
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000595
InChI Key OSOUNOBYRMOXQQ-UHFFFAOYSA-N
Synonym 3-chlorotoluene, m-chlorotoluene, benzene, 1-chloro-3-methyl, m-tolyl chloride, 3-chloro-1-methylbenzene, 1-methyl-3-chlorobenzene, toluene, m-chloro, meta-chlorotoluene, unii-5k8172k9tt, chlorotoluene 3-
PubChem CID 7931
IUPAC Name 1-chloro-3-methylbenzene
SMILES CC1=CC=CC(Cl)=C1

Specifications

Melting Point -48°C
Color Colorless to Yellow
CAS Min % 96.0
CAS Max % 100.0
Density 1.0700g/mL
Quantity 100 mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5210 to 1.523
Specific Gravity 1.07
Assay Percent Range 97%
Linear Formula CH3C6H4Cl
Beilstein 05, 291
Merck Index 15, 2175
Boiling Point 160°C to 162°C
Solubility Information Solubility in water: slightly soluble
Formula Weight 126.59
Percent Purity 97%
Chemical Name or Material 3-Chlorotoluene, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
EINECSNumber 203-580-5
TSCA TSCA

RUO – Research Use Only