missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorothiophene-2-carboxylic acid, 97+%

Catalog No. p-7046237
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1356003 1 g
AAL1356006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1356003 Supplier Thermo Scientific Chemicals Supplier No. L1356003
Only null left

CAS: 59337-89-2 | C5H3ClO2S | 162.587 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59337-89-2
Molecular Formula C5H3ClO2S
Molecular Weight (g/mol) 162.587
MDL Number MFCD00043888
InChI Key BXEAAHIHFFIMIE-UHFFFAOYSA-N
Synonym 3-chloro-2-thiophenecarboxylic acid, 3-chlorothiophene-2-carboxylicacid, 2-thiophenecarboxylic acid, chloro, 3-chloro-thiophene-2-carboxylic acid, pubchem5491, chlorothiophenecarboxylic, acmc-20agf8, maybridge1_004006, ksc274e4d, 3-chlorothiophenecarboxylic acid
PubChem CID 701269
IUPAC Name 3-chlorothiophene-2-carboxylic acid
SMILES C1=CSC(=C1Cl)C(=O)O

Specifications

Melting Point 185°C to 187°C
Quantity 1 g
Beilstein 121052
Formula Weight 162.6
Percent Purity 99%
Chemical Name or Material 3-Chlorothiophene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only