missing translation for 'onlineSavingsMsg'
Learn More

3-(Chlorosulfonyl)benzoic acid, 96%

Catalog No. p-7026777
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2298706 5 g
AAB2298714 25 g
AAB2298722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2298706 Supplier Thermo Scientific Chemicals Supplier No. B2298706
Only null left

CAS: 4025-64-3 | C7H5ClO4S | 220.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4025-64-3
Molecular Formula C7H5ClO4S
Molecular Weight (g/mol) 220.62
MDL Number MFCD00024877
InChI Key LMRKXSDOAFUINK-UHFFFAOYSA-N
Synonym 3-chlorosulfonyl benzoic acid, benzoic acid, 3-chlorosulfonyl, 3-carboxybenzenesulfonyl chloride, m-chlorosulfonylbenzoic acid, 3-carboxyphenylsulfonyl chloride, benzoic acid, m-chlorosulfonyl, 3-chlorosulphonyl benzoic acid, 3-carboxylbenzenesulfonyl chloride, 3-chlorosulfonyl-benzoic acid, m-chlorosulfonyl benzoic acid
PubChem CID 77641
SMILES OC(=O)C1=CC=CC(=C1)S(Cl)(=O)=O

Specifications

Melting Point 133°C to 138°C
Quantity 5 g
UN Number UN3261
Beilstein 2645465
Sensitivity Moisture sensitive
IUPAC Name 3-(chlorosulfonyl)benzoic acid
Formula Weight 220.63
Percent Purity 97%
Chemical Name or Material 3-(Chlorosulfonyl)benzoic acid
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 223-692-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only