missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorosalicylaldehyde 98.0+%, TCI America™

Catalog No. C31665G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31665G 5 g
C31661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31665G Supplier TCI America Supplier No. C31665G
Only null left

Chemical Identifiers

CAS 1927-94-2
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.565
MDL Number MFCD04973581
InChI Key DOHOPUBZLWVZMZ-UHFFFAOYSA-N
Synonym 3-Chloro-2-hydroxybenzaldehyde
PubChem CID 519651
IUPAC Name 3-chloro-2-hydroxybenzaldehyde
SMILES C1=CC(=C(C(=C1)Cl)O)C=O

Specifications

Melting Point 55°C
Boiling Point 63°C
Quantity 5 g
Formula Weight 156.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chlorosalicylaldehyde