missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorosalicylaldehyde 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 55°C |
| Boiling Point | 63°C |
| Quantity | 1 g |
| Formula Weight | 156.57 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-Chlorosalicylaldehyde |
Safety and Handling
| TSCA | No |