missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropyridine 95.0+%, TCI America™

Catalog No. p-7124330
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
500 g
Catalog No. Quantity
C028025G 25 g
C0280100G 100 g
C0280500G 500 g
3 options

Catalog No. C028025G

Supplier: TCI America C028025G

Only null left

Chemical Identifiers

CAS 626-60-8
Molecular Formula C5H4ClN
Molecular Weight (g/mol) 113.54
MDL Number MFCD00006375
InChI Key PWRBCZZQRRPXAB-UHFFFAOYSA-N
Synonym pyridine, 3-chloro, m-chloropyridine, unii-1m13huc1p4, ccris 1715, chloropyridine, b-chloropyridine, 3-chloro pyridine, 3-chloro-pyridine, 3-chloropyridine, 3-chloranylpyridine
PubChem CID 12287
IUPAC Name 3-chloropyridine
SMILES ClC1=CC=CN=C1

Specifications

Color Yellow
Boiling Point 148°C
Quantity 25 g
Formula Weight 113.54
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloropyridine
RTECSNumber US6125000
TSCA Yes