missing translation for 'onlineSavingsMsg'
Learn More

3-chloropyridine-2-carboxylic acid, 97%

Catalog No. AC429912500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429912500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC429912500 Supplier Thermo Scientific Chemicals Supplier No. 429912500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 57266-69-0 | C6H4ClNO2 | 157.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57266-69-0
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.56
MDL Number MFCD01646325
InChI Key XTMUXJBJCMRWPG-UHFFFAOYSA-N
Synonym 3-chloropicolinic acid, 3-chloro-2-pyridinecarboxylic acid, 3-chloro-2-picolinic acid, 2-pyridinecarboxylic acid, 3-chloro, 3-chloropyridine-2-carboxylicacid, 3-chloro-2-picolinicacid, 3-chloro-pyridine-2-carboxylic acid, chloropicolinic acid, pubchem12918, 3-chloro picolinic acid
PubChem CID 824632
IUPAC Name 3-chloropyridine-2-carboxylic acid
SMILES C1=CC(=C(N=C1)C(=O)O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 124.0°C to 126.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 157.56
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-chloropyridine-2-carboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only