missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropyrazine-2-carboxamide 98.0+%, TCI America™

Catalog No. p-7125945
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C3376200MG 200 mg
C33761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C3376200MG Supplier TCI America Supplier No. C3376200MG
Only null left

Chemical Identifiers

CAS 21279-62-9
Molecular Formula C5H4ClN3O
Molecular Weight (g/mol) 157.56
MDL Number MFCD09863915
InChI Key YHPMRHPLAQSPHJ-UHFFFAOYSA-N
PubChem CID 301266
IUPAC Name 3-chloropyrazine-2-carboxamide
SMILES NC(=O)C1=NC=CN=C1Cl

Specifications

Melting Point 189°C
Color White-Yellow
Quantity 200 mg
Formula Weight 157.56
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloropyrazine-2-carboxamide