missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloropropylamine Hydrochloride 97.0+%, TCI America™
Supplier: TCI America C106225G
Specifications
| 3-Chloropropylamine Hydrochloride | |
| 149°C | |
| C3H9Cl2N | |
| 25 g | |
| IHPRVZKJZGXTBQ-UHFFFAOYSA-N | |
| 3-chloropropan-1-amine;hydrochloride | |
| 11469095 | |
| ≥97.0% (T) |
| 6276-54-6 | |
| White-Yellow | |
| MFCD00012913 | |
| 3-chloropropylamine hydrochloride, 3-chloropropan-1-amine hydrochloride, 3-chloropropylamine hcl, 1-amino-3-chloropropane hydrochloride, 3-chloro-1-propanamine hydrochloride, 1-propanamine, 3-chloro-, hydrochloride, 3-chloropropylammonium chloride, acmc-1b7ws, chloropropylamine hydrochloride, ksc491e7b | |
| C(CN)CCl.Cl | |
| 130.012 | |
| 130.01 | |
| Crystalline Powder |
Chemical Identifiers
| 6276-54-6 | |
| 130.012 | |
| IHPRVZKJZGXTBQ-UHFFFAOYSA-N | |
| 11469095 | |
| C(CN)CCl.Cl |
| C3H9Cl2N | |
| MFCD00012913 | |
| 3-chloropropylamine hydrochloride, 3-chloropropan-1-amine hydrochloride, 3-chloropropylamine hcl, 1-amino-3-chloropropane hydrochloride, 3-chloro-1-propanamine hydrochloride, 1-propanamine, 3-chloro-, hydrochloride, 3-chloropropylammonium chloride, acmc-1b7ws, chloropropylamine hydrochloride, ksc491e7b | |
| 3-chloropropan-1-amine;hydrochloride |
Safety and Handling
TSCA : No