missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropylamine hydrochloride, 98%

Catalog No. AAA1541518
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1541518 50 g
AAA1541530 250 g
AAA154150B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1541518 Supplier Thermo Scientific Chemicals Supplier No. A1541518

CAS: 6276-54-6 | C3H9Cl2N | 130.012 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6276-54-6
Molecular Formula C3H9Cl2N
Molecular Weight (g/mol) 130.012
MDL Number MFCD00012913
InChI Key IHPRVZKJZGXTBQ-UHFFFAOYSA-N
Synonym 3-chloropropylamine hydrochloride, 3-chloropropan-1-amine hydrochloride, 3-chloropropylamine hcl, 1-amino-3-chloropropane hydrochloride, 3-chloro-1-propanamine hydrochloride, 1-propanamine, 3-chloro-, hydrochloride, 3-chloropropylammonium chloride, acmc-1b7ws, chloropropylamine hydrochloride, ksc491e7b
PubChem CID 11469095
IUPAC Name 3-chloropropan-1-amine;hydrochloride
SMILES C(CN)CCl.Cl

Specifications

Melting Point 147°C to 152°C
Quantity 50 g
Beilstein 3667451
Formula Weight 130.02
Percent Purity 98%
Chemical Name or Material 3-Chloropropylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-474-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only