missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropylamine hydrochloride, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 162010010

Quantity 1kg
Packaging Plastic bottle

 View more versions of this product

Catalog No. AC162010010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

6276-54-6
130.02
IHPRVZKJZGXTBQ-UHFFFAOYSA-N
11469095
C(CN)CCl.Cl
C3H8ClN·HCl
MFCD00012913
3-chloropropylamine hydrochloride, 3-chloropropan-1-amine hydrochloride, 3-chloropropylamine hcl, 1-amino-3-chloropropane hydrochloride, 3-chloro-1-propanamine hydrochloride, 1-propanamine, 3-chloro-, hydrochloride, 3-chloropropylammonium chloride, acmc-1b7ws, chloropropylamine hydrochloride, ksc491e7b
3-chloropropan-1-amine;hydrochloride

Specifications

3-Chloropropylamine hydrochloride
6276-54-6
100.0
98% min. (Ionised Chloride) (Argentometry)
C3H8ClN·HCl
1kg
04,148
Solubility in water: freely soluble
C(CN)CCl.Cl
130.02
130.02
98%
97.5
Authentic
Plastic bottle
Cl(CH2)3NH2·HCl
MFCD00012913
3-chloropropylamine hydrochloride, 3-chloropropan-1-amine hydrochloride, 3-chloropropylamine hcl, 1-amino-3-chloropropane hydrochloride, 3-chloro-1-propanamine hydrochloride, 1-propanamine, 3-chloro-, hydrochloride, 3-chloropropylammonium chloride, acmc-1b7ws, chloropropylamine hydrochloride, ksc491e7b
IHPRVZKJZGXTBQ-UHFFFAOYSA-N
3-chloropropan-1-amine;hydrochloride
11469095
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-474-6

TSCA : TSCA

SDS
Documents

Documents