missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropiophenone 96.0+%, TCI America™

Catalog No. C099525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C099525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C099525G Supplier TCI America Supplier No. C099525G

Chemical Identifiers

CAS 936-59-4
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00000990
InChI Key KTJRGPZVSKWRTJ-UHFFFAOYSA-N
Synonym 3-chloropropiophenone, 1-propanone, 3-chloro-1-phenyl, beta-chloropropiophenone, 2-chloroethyl phenyl ketone, 3-chloropropanophenone, 3-chloro-1-phenyl-1-propanone, propiophenone, 3-chloro, 3-chloro-1-phenyl-propan-1-one, .beta.-chloropropiophenone, .omega.-chloropropiophenone
PubChem CID 70295
IUPAC Name 3-chloro-1-phenylpropan-1-one
SMILES C1=CC=C(C=C1)C(=O)CCCl

Specifications

Melting Point 48°C
Color White-Yellow
Boiling Point 147°C
Quantity 25 g
Formula Weight 168.62
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloropropiophenone