missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloropropiophenone, 98%

Catalog No. AAA1254236
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1254236 500 g
AAA1254214 25 g
AAA1254222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1254236 Supplier Thermo Scientific Chemicals Supplier No. A1254236
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 34841-35-5 | C9H9ClO | 168.62 g/mol

3'-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′-Chloropropiophenone has been used in the preparation of 1-(3-chlorophenyl)-1-phenyl-1-propanol.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 34841-35-5
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00009925
InChI Key PQWGFUFROKIJBO-UHFFFAOYSA-N
Synonym 3'-chloropropiophenone, 1-3-chlorophenyl propan-1-one, m-chloropropiophenone, 3-chlorophenyl ethyl ketone, 1-propanone, 1-3-chlorophenyl, 1-3-chloro-phenyl-propan-1-one, m-chloro-propiophenone, pubchem3083, 3'-chloro-propiophenone, 3/'-chloropropiophenone
PubChem CID 587128
IUPAC Name 1-(3-chlorophenyl)propan-1-one
SMILES CCC(=O)C1=CC(=CC=C1)Cl

Specifications

Melting Point 45°C to 47°C
Boiling Point 123°C to 124°C (13 mmHg)
Flash Point >110°C (230°F)
Quantity 500 g
Solubility Information Soluble in methanol.
Formula Weight 168.62
Percent Purity 98%
Chemical Name or Material 3'-Chloropropiophenone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 252-242-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only