missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloropropiophenone, 98%

Catalog No. p-91440
Change view
Click to view available options
Quantity:
25 g
250 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC276580250 25 g Glass bottle
AC276582500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC276580250 Supplier Thermo Scientific Chemicals Supplier No. 276580250

CAS: 34841-35-5 | C9H9ClO | 168.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 34841-35-5
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00009925
InChI Key PQWGFUFROKIJBO-UHFFFAOYSA-N
Synonym 3'-chloropropiophenone, 1-3-chlorophenyl propan-1-one, m-chloropropiophenone, 3-chlorophenyl ethyl ketone, 1-propanone, 1-3-chlorophenyl, 1-3-chloro-phenyl-propan-1-one, m-chloro-propiophenone, pubchem3083, 3'-chloro-propiophenone, 3/'-chloropropiophenone
PubChem CID 587128
IUPAC Name 1-(3-chlorophenyl)propan-1-one
SMILES CCC(=O)C1=CC(=CC=C1)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 43°C to 47°C
Color White to Yellow
Boiling Point 124°C (14.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClC6H4COC2H5
Quantity 25 g
Beilstein 07, 1028
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in hot toluene and alcohol
Formula Weight 168.62
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 3'-Chloropropiophenone, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 252-242-3
TSCA TSCA

RUO – Research Use Only