missing translation for 'onlineSavingsMsg'
Learn More

3-Chloropropanesulfonyl chloride, 98+%

Catalog No. AAA1063109
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1063109 10 g
AAA1063118 50 g
AAA1063130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1063109 Supplier Thermo Scientific Chemicals Supplier No. A1063109
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1633-82-5 | C3H6Cl2O2S | 177.039 g/mol

3-Chloropropanesulfonyl Chloride is an intermediate used in the preparation of many enzymatic inhibitors. It is also used in the generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids provide chiral sultams and sultones. It is mainly used for the synthesis of pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloropropanesulfonyl Chloride is an intermediate used in the preparation of many enzymatic inhibitors. It is also used in the generation and trapping of the derived sulfene with imines and glyoxylates in the presence of chinchona alkaloids provide chiral sultams and sultones. It is mainly used for the synthesis of pharmaceutical intermediates.

Solubility
Soluble in dichloromethane, ethyl Acetate and hexane.

Notes
Store at -20°C. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Moisture sensitive. Keep away from alcohols, strong bases, oxidizing agents.

Chemical Identifiers

CAS 1633-82-5
Molecular Formula C3H6Cl2O2S
Molecular Weight (g/mol) 177.039
MDL Number MFCD00007463
InChI Key GPKDGVXBXQTHRY-UHFFFAOYSA-N
Synonym 3-chloropropanesulfonyl chloride, 3-chloro-1-propanesulfonyl chloride, 3-chloropropanesulfonylchloride, 3-chloropropylsulfonyl chloride, 3-chloropropanesulphonyl chloride, 1-propanesulfonyl chloride, 3-chloro, propanesulfonyl chloride, chloro, 3-chloro-propane-1-sulfonyl chloride, gamma-chloropropanesulfonyl chloride, 3-chloropropylsulfonylchloride
PubChem CID 15410
IUPAC Name 3-chloropropane-1-sulfonyl chloride
SMILES C(CS(=O)(=O)Cl)CCl

Specifications

Density 1.456
Boiling Point 70°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.49
Linear Formula Cl(CH2)3SO2Cl
Quantity 10 g
UN Number UN3265
Beilstein 1754119
Sensitivity Moisture sensitive
Solubility Information Soluble in dichloromethane,ethyl Acetate and hexane.
Formula Weight 177.05
Percent Purity ≥98%
Chemical Name or Material 3-Chloropropanesulfonyl chloride
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 216-646-3
RTECSNumber TZ6670000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only