missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophthalic Anhydride 98.0+%, TCI America™

Catalog No. p-7125973
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34895G 5 g
C348925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34895G Supplier TCI America Supplier No. C34895G
Only null left

Chemical Identifiers

CAS 117-21-5
Molecular Formula C8H3ClO3
Molecular Weight (g/mol) 182.56
MDL Number MFCD00023107
InChI Key UERPUZBSSSAZJE-UHFFFAOYSA-N
Synonym 3-chlorophthalic anhydride, 4-chloroisobenzofuran-1,3-dione, 3-chlorophthalic acid anhydride, unii-vo7nxt0137, 1,3-isobenzofurandione, chloro, 4-chloro-1,3-dihydro-2-benzofuran-1,3-dione, 4-chloranyl-2-benzofuran-1,3-dione, 3clpa, pubchem19464, 3-chlorophthalicanhydride
PubChem CID 67014
IUPAC Name 4-chloro-1,3-dihydro-2-benzofuran-1,3-dione
SMILES ClC1=CC=CC2=C1C(=O)OC2=O

Specifications

Melting Point 128°C
Color White
Boiling Point 170°C
Quantity 5 g
Formula Weight 182.56
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chlorophthalic Anhydride
EINECSNumber (3)-0062
TSCA No