missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$67.11 - $67.11
Specifications
| CAS | 63503-60-6 |
|---|---|
| Physical Form | Crystal-Powder at 20°C |
| Molecular Formula | C6H6BClO2 |
| MDL Number | MFCD00161354 |
| InChI Key | SDEAGACSNFSZCU-UHFFFAOYSA-N |
Specifications
| 63503-60-6 | |
| C6H6BClO2 | |
| SDEAGACSNFSZCU-UHFFFAOYSA-N | |
| 2734323 |
| Crystal-Powder at 20°C | |
| MFCD00161354 | |
| (3-chlorophenyl)boronic acid | |
| 156.37 |