missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. C16135G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C16135G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C16135G Supplier TCI America Supplier No. C16135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Chlorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 63503-60-6
Color White-Yellow
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Molecular Formula C6H6BClO2
MDL Number MFCD00161354
Synonym 3-chlorophenyl boronic acid, 3-chlorobenzeneboronic acid, 3-chlorophenyl boranediol, 3-chlorophenylbornic acid, 3-chloro benzene boronic acid, m-chlorophenylboronic acid, 3-chloro-phenyl-boronic acid, boronic acid, 3-chlorophenyl, pubchem1785
InChI Key SDEAGACSNFSZCU-UHFFFAOYSA-N
SMILES OB(O)C1=CC(Cl)=CC=C1
IUPAC Name (3-chlorophenyl)boronic acid
Molecular Weight (g/mol) 156.37
PubChem CID 2734323
Formula Weight 156.37
Show More Show Less