Learn More
3-Chlorophenylboronic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 63503-60-6 |
|---|---|
| Molecular Formula | C6H6BClO2 |
| Molecular Weight (g/mol) | 156.38 |
| MDL Number | MFCD00161354 |
| InChI Key | SDEAGACSNFSZCU-UHFFFAOYSA-N |
| Synonym | 3-chlorophenyl boronic acid, 3-chlorobenzeneboronic acid, 3-chlorophenyl boranediol, 3-chlorophenylbornic acid, 3-chloro benzene boronic acid, m-chlorophenylboronic acid, 3-chloro-phenyl-boronic acid, boronic acid, 3-chlorophenyl, pubchem1785 |
| PubChem CID | 2734323 |
| IUPAC Name | (3-chlorophenyl)boronic acid |
| SMILES | B(C1=CC(=CC=C1)Cl)(O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC359000010
|
Thermo Scientific Chemicals
359000010 |
1g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC359000050
|
Thermo Scientific Chemicals
359000050 |
5g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 63503-60-6 | |
| 156.38 | |
| SDEAGACSNFSZCU-UHFFFAOYSA-N | |
| 2734323 | |
| B(C1=CC(=CC=C1)Cl)(O)O |
| C6H6BClO2 | |
| MFCD00161354 | |
| 3-chlorophenyl boronic acid, 3-chlorobenzeneboronic acid, 3-chlorophenyl boranediol, 3-chlorophenylbornic acid, 3-chloro benzene boronic acid, m-chlorophenylboronic acid, 3-chloro-phenyl-boronic acid, boronic acid, 3-chlorophenyl, pubchem1785 | |
| (3-chlorophenyl)boronic acid |
Specifications
| 63503-60-6 | |
| 100.0 | |
| 96% min. (HPLC) | |
| C6H6BClO2 | |
| 1g | |
| SDEAGACSNFSZCU-UHFFFAOYSA-N | |
| (3-chlorophenyl)boronic acid | |
| 2734323 | |
| 97% |
| 96.0 | |
| Authentic | |
| Glass bottle | |
| MFCD00161354 | |
| 3-chlorophenyl boronic acid, 3-chlorobenzeneboronic acid, 3-chlorophenyl boranediol, 3-chlorophenylbornic acid, 3-chloro benzene boronic acid, m-chlorophenylboronic acid, 3-chloro-phenyl-boronic acid, boronic acid, 3-chlorophenyl, pubchem1785 | |
| B(C1=CC(=CC=C1)Cl)(O)O | |
| 156.38 | |
| 156.38 | |
| 3-Chlorophenylboronic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
RUO â Research Use Only