missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenyl isothiocyanate, 97%

CAS: 2392-68-9 | C7H4ClNS | 169.63 g/mol

$74.51 - $229.49

Chemical Identifiers

CAS 2392-68-9
Molecular Formula C7H4ClNS
Molecular Weight (g/mol) 169.63
MDL Number MFCD00004805
InChI Key WGXCKFMVBAOIFH-UHFFFAOYSA-N
Synonym 3-chlorophenyl isothiocyanate, 3-chlorophenylisothiocyanate, benzene, 1-chloro-3-isothiocyanato, isothiocyanic acid, 3-chlorophenyl ester, isothiocyanic acid, m-chlorophenyl ester, m-chlorophenyl isothiocyanate, 1-chloro-3-isothiocyanato-benzene, 3-chlorobenzenisothiocyanate, wln: scnr cg, acmc-1cfb8
PubChem CID 75449
IUPAC Name 1-chloro-3-isothiocyanatobenzene
SMILES ClC1=CC=CC(=C1)N=C=S
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1199906
View Documents
Thermo Scientific Chemicals
A1199906
5 g
Each for $74.51
Only null left
 
AAA1199914
View Documents
Thermo Scientific Chemicals
A1199914
25 g
Each for $229.49
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2392-68-9
169.63
WGXCKFMVBAOIFH-UHFFFAOYSA-N
75449
ClC1=CC=CC(=C1)N=C=S
C7H4ClNS
MFCD00004805
3-chlorophenyl isothiocyanate, 3-chlorophenylisothiocyanate, benzene, 1-chloro-3-isothiocyanato, isothiocyanic acid, 3-chlorophenyl ester, isothiocyanic acid, m-chlorophenyl ester, m-chlorophenyl isothiocyanate, 1-chloro-3-isothiocyanato-benzene, 3-chlorobenzenisothiocyanate, wln: scnr cg, acmc-1cfb8
1-chloro-3-isothiocyanatobenzene

Specifications

2392-68-9
249°C to 250°C
Pungent
C7H4ClNS
UN2922
3-chlorophenyl isothiocyanate, 3-chlorophenylisothiocyanate, benzene, 1-chloro-3-isothiocyanato, isothiocyanic acid, 3-chlorophenyl ester, isothiocyanic acid, m-chlorophenyl ester, m-chlorophenyl isothiocyanate, 1-chloro-3-isothiocyanato-benzene, 3-chlorobenzenisothiocyanate, wln: scnr cg, acmc-1cfb8
ClC1=CC=CC(=C1)N=C=S
169.63
169.64
3-Chlorophenyl isothiocyanate
1.281
5 g
1.6585
MFCD00004805
636856
WGXCKFMVBAOIFH-UHFFFAOYSA-N
1-chloro-3-isothiocyanatobenzene
75449
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c

H301+H311+H331-H315-H319-H334-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: II

EINECSNumber : 000-000-0

RTECSNumber : NX8474500

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only