missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)pyridine Hydrochloride 98.0+%, TCI America™

Catalog No. p-7124631
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C082525G 25 g
C0825100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C082525G Supplier TCI America Supplier No. C082525G
Only null left

Chemical Identifiers

CAS 6959-48-4
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00012818
InChI Key UZGLOGCJCWBBIV-UHFFFAOYSA-N
Synonym 3-chloromethyl pyridine hydrochloride, 3-chloromethylpyridine hydrochloride, 3-picolyl chloride hydrochloride, 3-chloromethyl pyridine hcl, ccris 140, pyridine, 3-chloromethyl-, hydrochloride, 3-pyridylmethyl chloride hydrochloride, unii-bqa8fyp4yn, 3-chloromethyl pyridinium chloride, 3-chloromethyl pyridine.hcl
PubChem CID 23394
IUPAC Name 3-(chloromethyl)pyridine;hydrochloride
SMILES C1=CC(=CN=C1)CCl.Cl

Specifications

Melting Point 144°C
Color White
Quantity 25 g
UN Number 3261
Formula Weight 164.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-(Chloromethyl)pyridine Hydrochloride
RTECSNumber US7000000
TSCA No