missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Chloromethyl)pyridine Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 144°C |
| Color | White |
| Quantity | 25 g |
| UN Number | 3261 |
| Formula Weight | 164.03 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 3-(Chloromethyl)pyridine Hydrochloride |
Safety and Handling
| RTECSNumber | US7000000 |
| TSCA | No |