missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)pyridine hydrochloride, 97%

Catalog No. AAA1327906
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1327906 5 g
AAA1327914 25 g
AAA1327922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1327906 Supplier Thermo Scientific Chemicals Supplier No. A1327906

CAS: 6959-48-4 | C6H7Cl2N | 164.029 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6959-48-4
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00012818
InChI Key UZGLOGCJCWBBIV-UHFFFAOYSA-N
Synonym 3-chloromethyl pyridine hydrochloride, 3-chloromethylpyridine hydrochloride, 3-picolyl chloride hydrochloride, 3-chloromethyl pyridine hcl, ccris 140, pyridine, 3-chloromethyl-, hydrochloride, 3-pyridylmethyl chloride hydrochloride, unii-bqa8fyp4yn, 3-chloromethyl pyridinium chloride, 3-chloromethyl pyridine.hcl
PubChem CID 23394
IUPAC Name 3-(chloromethyl)pyridine;hydrochloride
SMILES C1=CC(=CN=C1)CCl.Cl

Specifications

Melting Point 136°C to 142°C
pH 3
Quantity 5 g
UN Number UN3261
Beilstein 3689156
Sensitivity Hygroscopic
Formula Weight 164.04
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)pyridine hydrochloride
Hazard Category H302-H314
Hazard Statement GHS H Statement
H301-H311-H314-H318-H341-H351-H317
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of causing genetic defects.
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 230-150-4
RTECSNumber US7000000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only