missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorocinnamic Acid 98.0+%, TCI America™

Catalog No. C097525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C097525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C097525G Supplier TCI America Supplier No. C097525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1866-38-2
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.603
MDL Number MFCD00004384
InChI Key FFKGOJWPSXRALK-SNAWJCMRSA-N
Synonym 3-chlorocinnamic acid, 3-3-chlorophenyl acrylic acid, m-chlorocinnamic acid, e-3-3-chlorophenyl acrylic acid, 2e-3-3-chlorophenyl acrylic acid, trans-3-chlorocinnamic acid, e-3-chlorocinnamic acid, 2e-3-3-chlorophenyl prop-2-enoic acid, 2-propenoic acid, 3-3-chlorophenyl, rarechem bk hc t320
PubChem CID 735250
IUPAC Name (E)-3-(3-chlorophenyl)prop-2-enoic acid
SMILES C1=CC(=CC(=C1)Cl)C=CC(=O)O

Specifications

Melting Point 164°C
Color White-Yellow
Quantity 25 g
Formula Weight 182.60
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chlorocinnamic Acid