missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorocarbonylbenzeneboronic anhydride, tech. 90%

Catalog No. AAH5272203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5272203 1 g
AAH5272206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5272203 Supplier Thermo Scientific Chemicals Supplier No. H5272203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 332154-58-2 | C7H6BClO3 | 184.382 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 332154-58-2
Molecular Formula C7H6BClO3
Molecular Weight (g/mol) 184.382
MDL Number MFCD06801675
InChI Key AIHGXUQQIIYAJP-UHFFFAOYSA-N
Synonym 3-chlorocarbonylphenylboronic acid, 3-chlorocarbonyl phenyl boronic acid, benzoylchloride-3-boronic acid, 3-carbonochloridoyl phenylboronic acid, pubchem9524, acmc-209hzb, 3-chlorocarbonylphenylboronicacid, boronic acid, 3-chlorocarbonyl phenyl-9ci
PubChem CID 17750521
IUPAC Name (3-carbonochloridoylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)C(=O)Cl)(O)O

Specifications

Melting Point 252°C to 254°C
Quantity 1 g
Formula Weight 166.37
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Chlorocarbonylbenzeneboronic anhydride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only