missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzo[b]thiophene-2-carboxylic acid, 97%

CAS: 21211-22-3 | C9H5ClO2S | 212.647 g/mol

$71.33 - $248.90

Chemical Identifiers

CAS 21211-22-3
Molecular Formula C9H5ClO2S
Molecular Weight (g/mol) 212.647
MDL Number MFCD00014579
InChI Key HJTMIYKPPPYDRJ-UHFFFAOYSA-N
Synonym 3-chlorobenzo b thiophene-2-carboxylic acid, 3-chloro-benzo b thiophene-2-carboxylic acid, 3-chloro-benzo b thiophene-2-carboxylicacid, benzo b thiophene-2-carboxylic acid, 3-chloro, 3-chlorobenzothiophene-2-carboxylic acid, pubchem18541, 3-chloro-benzo b thiophene carboxylic acid, acmc-1coc0, cbmicro_009255
PubChem CID 307964
IUPAC Name 3-chloro-1-benzothiophene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=C(S2)C(=O)O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0125703
View Documents
Thermo Scientific Chemicals
L0125703
1 g
Each for $71.33
Only null left
 
AAL0125706
View Documents
Thermo Scientific Chemicals
L0125706
5 g
Each for $248.90
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

21211-22-3
212.647
HJTMIYKPPPYDRJ-UHFFFAOYSA-N
307964
C1=CC=C2C(=C1)C(=C(S2)C(=O)O)Cl
C9H5ClO2S
MFCD00014579
3-chlorobenzo b thiophene-2-carboxylic acid, 3-chloro-benzo b thiophene-2-carboxylic acid, 3-chloro-benzo b thiophene-2-carboxylicacid, benzo b thiophene-2-carboxylic acid, 3-chloro, 3-chlorobenzothiophene-2-carboxylic acid, pubchem18541, 3-chloro-benzo b thiophene carboxylic acid, acmc-1coc0, cbmicro_009255
3-chloro-1-benzothiophene-2-carboxylic acid

Specifications

21211-22-3
C9H5ClO2S
1 g
3-chlorobenzo b thiophene-2-carboxylic acid, 3-chloro-benzo b thiophene-2-carboxylic acid, 3-chloro-benzo b thiophene-2-carboxylicacid, benzo b thiophene-2-carboxylic acid, 3-chloro, 3-chlorobenzothiophene-2-carboxylic acid, pubchem18541, 3-chloro-benzo b thiophene carboxylic acid, acmc-1coc0, cbmicro_009255
C1=CC=C2C(=C1)C(=C(S2)C(=O)O)Cl
212.647
212.65
3-Chlorobenzo[b]thiophene-2-carboxylic acid
265°C to 267°C
MFCD00014579
1425602
HJTMIYKPPPYDRJ-UHFFFAOYSA-N
3-chloro-1-benzothiophene-2-carboxylic acid
307964
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only