missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzo[b]thiophene-2-carboxylic acid, 97%

Catalog No. AAL0125703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0125703 1 g
AAL0125706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0125703 Supplier Thermo Scientific Chemicals Supplier No. L0125703

CAS: 21211-22-3 | C9H5ClO2S | 212.647 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21211-22-3
Molecular Formula C9H5ClO2S
Molecular Weight (g/mol) 212.647
MDL Number MFCD00014579
InChI Key HJTMIYKPPPYDRJ-UHFFFAOYSA-N
Synonym 3-chlorobenzo b thiophene-2-carboxylic acid, 3-chloro-benzo b thiophene-2-carboxylic acid, 3-chloro-benzo b thiophene-2-carboxylicacid, benzo b thiophene-2-carboxylic acid, 3-chloro, 3-chlorobenzothiophene-2-carboxylic acid, pubchem18541, 3-chloro-benzo b thiophene carboxylic acid, acmc-1coc0, cbmicro_009255
PubChem CID 307964
IUPAC Name 3-chloro-1-benzothiophene-2-carboxylic acid
SMILES C1=CC=C2C(=C1)C(=C(S2)C(=O)O)Cl

Specifications

Melting Point 265°C to 267°C
Quantity 1 g
Beilstein 1425602
Formula Weight 212.65
Percent Purity 97%
Chemical Name or Material 3-Chlorobenzo[b]thiophene-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only