missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzo[b]thiophene-2-carbonyl chloride, 95%

Catalog No. p-7045721
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1174503 1 g
AAL1174506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1174503 Supplier Thermo Scientific Chemicals Supplier No. L1174503
Only null left

CAS: 21815-91-8 | C9H4Cl2OS | 231.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21815-91-8
Molecular Formula C9H4Cl2OS
Molecular Weight (g/mol) 231.09
MDL Number MFCD00053069
InChI Key GWKSSMDJEWPKCM-UHFFFAOYSA-N
Synonym 3-chlorobenzo b thiophene-2-carbonyl chloride, 3-chlorobenzothiophene-2-carbonyl chloride, 3-chloro-benzo b thiophene-2-carbonyl chloride, 3-chlorobenzo b thiophen-2-carbonyl chloride, benzo b thiophene-2-carbonyl chloride, 3-chloro, 3-chlorobenzo b-2-thiophenecarboxylic acid chloride, acmc-1cne6, 3-chlorobenzo b thiophene-2-carbonylchloride, 2-chlorocarbonyl-3-chlorobenzo b thiophene, 3-chlorobenzo thiophene-2-carbonyl chloride
PubChem CID 519898
IUPAC Name 3-chloro-1-benzothiophene-2-carbonyl chloride
SMILES ClC(=O)C1=C(Cl)C2=CC=CC=C2S1

Specifications

Melting Point 111°C to 114°C
Quantity 1 g
UN Number UN3261
Beilstein 1309703
Sensitivity Moisture sensitive
Formula Weight 231.1
Percent Purity 95%
Chemical Name or Material 3-Chlorobenzo[b]thiophene-2-carbonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 625-793-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only