missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chlorobenzenesulfonyl Chloride 98.0+%, TCI America™
Supplier: TCI America C17911G
Specifications
| 3-Chlorobenzenesulfonyl Chloride | |
| White-Yellow | |
| C6H4Cl2O2S | |
| 1 g | |
| 3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4b | |
| C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl | |
| 211.06 | |
| 211.06 | |
| Crystalline Lumps |
| 2888-06-4 | |
| 135°C | |
| MFCD00051697 | |
| 3261 | |
| OINWZUJVEXUHCC-UHFFFAOYSA-N | |
| 3-chlorobenzenesulfonyl chloride | |
| 17909 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 2888-06-4 | |
| 211.06 | |
| OINWZUJVEXUHCC-UHFFFAOYSA-N | |
| 17909 | |
| C1=CC(=CC(=C1)Cl)S(=O)(=O)Cl |
| C6H4Cl2O2S | |
| MFCD00051697 | |
| 3-chlorobenzene-1-sulfonyl chloride, 3-chlorobenzenesulphonyl chloride, 3-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 3-chloro, m-chlorobenzenesulfonyl chloride, 3-chloro-benzenesulfonyl chloride, 3-chlorophenylsulfonyl chloride, pubchem5076, m-chlorbenzolsulfochlorid, acmc-1cj4b | |
| 3-chlorobenzenesulfonyl chloride |
Safety and Handling
TSCA : No