missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzeneboronic acid, 97%

Catalog No. AAB2444403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2444403 1 g
AAB2444406 5 g
AAB2444414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2444403 Supplier Thermo Scientific Chemicals Supplier No. B2444403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 63503-60-6 | C6H6BClO2 | 156.37 g/mol

It is employed as a reactant involved in 1,4-conjugate addition reactions with ethenesulfonamides to form arylethanesulfonamides, Suzuki-Miyaura reactions with dibromotrifluoromethylbenzene, cross-coupling reactions with diazoesters3 or potassium cyanate. It is involved in the synthesis of biarylketones and phthalides and synthesis of inhibitors including PDE4 inhibitors6 among others.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reactant involved in 1,4-conjugate addition reactions with ethenesulfonamides to form arylethanesulfonamides, Suzuki-Miyaura reactions with dibromotrifluoromethylbenzene, cross-coupling reactions with diazoesters3 or potassium cyanate. It is involved in the synthesis of biarylketones and phthalides and synthesis of inhibitors including PDE4 inhibitors6 among others.

Solubility
Slightly soluble in water. soluble in ether, tetrahydrofuran,, dimethyl sulfoxide, dimethyl formamide, methanol.

Notes
Store at room temperature. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 63503-60-6
Molecular Formula C6H6BClO2
Molecular Weight (g/mol) 156.37
MDL Number MFCD00161354
InChI Key SDEAGACSNFSZCU-UHFFFAOYSA-N
Synonym 3-chlorophenyl boronic acid, 3-chlorobenzeneboronic acid, 3-chlorophenyl boranediol, 3-chlorophenylbornic acid, 3-chloro benzene boronic acid, m-chlorophenylboronic acid, 3-chloro-phenyl-boronic acid, boronic acid, 3-chlorophenyl, pubchem1785
PubChem CID 2734323
IUPAC Name (3-chlorophenyl)boronic acid
SMILES OB(O)C1=CC(Cl)=CC=C1

Specifications

Melting Point 180°C to 185°C
Quantity 1 g
Beilstein 2936343
Solubility Information Slightly soluble in water. soluble in ether,tetrahydrofuran,,dimethyl sulfoxide,dimethyl formamide,methanol.
Formula Weight 156.38
Percent Purity 97%
Chemical Name or Material 3-Chlorobenzeneboronic acid
Hazard Category H302-H312-H315-H319-H332-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only