missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzamide, 98+%, Thermo Scientific Chemicals

Catalog No. p-7081419
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1138909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1138909 Supplier Thermo Scientific Chemicals Supplier No. A1138909

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 618-48-4 | Molecular Formula: C7H6ClNO | Molecular Weight: 155.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 618-48-4
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD00014799
InChI Key MJTGQALMWUUPQM-UHFFFAOYSA-N
Synonym m-chlorobenzamide, benzamide, 3-chloro, 3-chloro-benzamide, benzamide, m-chloro-8ci, benzamide, m-chloro, 5-chlorobenzamide, pubchem3618, acmc-1b3cu, wce
PubChem CID 69254
ChEBI CHEBI:10587
IUPAC Name 3-chlorobenzamide
SMILES NC(=O)C1=CC=CC(Cl)=C1

Specifications

Melting Point 133°C to 136°C
Quantity 10 g
Beilstein 1859940
Formula Weight 155.58
Percent Purity ≥98%
Chemical Name or Material 3-Chlorobenzamide
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-554-7
RTECSNumber CV2443774
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only