missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroaniline Hydrochloride 99.0+%, TCI America™

Catalog No. C011325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C011325G 25 g
C0113500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C011325G Supplier TCI America Supplier No. C011325G
Only null left

Chemical Identifiers

CAS 141-85-5
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.029
MDL Number MFCD00035485
InChI Key NMGHWHCTRGZZOP-UHFFFAOYSA-N
Synonym 3-chloroaniline hydrochloride, 3-chloroanilinium chloride, devol orange c, devol orange gc, m-chloroaniline hydrochloride, orange gc salt, orange base ngc, orange gcs salt, orange salt ngc, ansibases orange gc
PubChem CID 67336
IUPAC Name 3-chloroaniline;hydrochloride
SMILES C1=CC(=CC(=C1)Cl)N.Cl

Specifications

Quantity 25 g
Formula Weight 164.03
Percent Purity ≥99.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloroaniline Hydrochloride
RTECSNumber BX1393800
TSCA Yes