missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloroacetophenone 97.0+%, TCI America™

Catalog No. p-7124214
Change view
Click to view available options
Quantity:
10 mL
25 mL
Catalog No. Quantity
C009610ML 10 mL
C009625ML 25 mL
2 options

Catalog No. C009610ML

Supplier: TCI America C009610ML

Only null left

Chemical Identifiers

CAS 99-02-5
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000593
InChI Key UUWJBXKHMMQDED-UHFFFAOYSA-N
Synonym 3'-chloroacetophenone, 1-3-chlorophenyl ethanone, m-chloroacetophenone, ethanone, 1-3-chlorophenyl, 3-chloroacetophenone, 1-3-chlorophenyl ethan-1-one, 3-chloro-acetophenone, acetophenone, 3'-chloro, 1-3-chloro-phenyl-ethanone, 3/'-chloroacetophenone
PubChem CID 14933
IUPAC Name 1-(3-chlorophenyl)ethan-1-one
SMILES CC(=O)C1=CC=CC(Cl)=C1

Specifications

Color Yellow
Boiling Point 230°C
Quantity 10 mL
UN Number 3416
Formula Weight 154.59
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3′-Chloroacetophenone
EINECSNumber (3)-1241
TSCA Yes