missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloroacetophenone 97.0+%, TCI America™

Catalog No. C009610ML
Change view
Click to view available options
Quantity:
10 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C009610ML 10 mL
C009625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C009610ML Supplier TCI America Supplier No. C009610ML

Chemical Identifiers

CAS 99-02-5
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
MDL Number MFCD00000593
InChI Key UUWJBXKHMMQDED-UHFFFAOYSA-N
Synonym 3'-chloroacetophenone, 1-3-chlorophenyl ethanone, m-chloroacetophenone, ethanone, 1-3-chlorophenyl, 3-chloroacetophenone, 1-3-chlorophenyl ethan-1-one, 3-chloro-acetophenone, acetophenone, 3'-chloro, 1-3-chloro-phenyl-ethanone, 3/'-chloroacetophenone
PubChem CID 14933
IUPAC Name 1-(3-chlorophenyl)ethan-1-one
SMILES CC(=O)C1=CC=CC(Cl)=C1

Specifications

Color Yellow
Boiling Point 230°C
Quantity 10 mL
UN Number 3416
Formula Weight 154.59
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3′-Chloroacetophenone
EINECSNumber (3)-1241
TSCA Yes