missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloroacetophenone, 98+%

CAS: 99-02-5 | C8H7ClO | 154.59 g/mol

Supplier:  Thermo Scientific Chemicals A1559214

 View more versions of this product

Catalog No. AAA1559214


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3'-Chloroacetophenone
1.191
105°C (221°F)
1.5506
25 g
636318
UUWJBXKHMMQDED-UHFFFAOYSA-N
1-(3-chlorophenyl)ethanone
14933
≥98%
99-02-5
227°C to 229°C
C8H7ClO
MFCD00000593
UN3416
3'-chloroacetophenone, 1-3-chlorophenyl ethanone, m-chloroacetophenone, ethanone, 1-3-chlorophenyl, 3-chloroacetophenone, 1-3-chlorophenyl ethan-1-one, 3-chloro-acetophenone, acetophenone, 3'-chloro, 1-3-chloro-phenyl-ethanone, 3/'-chloroacetophenone
CC(=O)C1=CC=CC(Cl)=C1
154.59
154.6

Chemical Identifiers

99-02-5
154.59
UUWJBXKHMMQDED-UHFFFAOYSA-N
14933
CC(=O)C1=CC=CC(Cl)=C1
C8H7ClO
MFCD00000593
3'-chloroacetophenone, 1-3-chlorophenyl ethanone, m-chloroacetophenone, ethanone, 1-3-chlorophenyl, 3-chloroacetophenone, 1-3-chlorophenyl ethan-1-one, 3-chloro-acetophenone, acetophenone, 3'-chloro, 1-3-chloro-phenyl-ethanone, 3/'-chloroacetophenone
1-(3-chlorophenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P362-P501c

H302-H315-H319-H331-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROACETOPHENONE, LIQUID

EINECSNumber : 202-721-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only