missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-o-xylene 96.0+%, TCI America™

Catalog No. c155825g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
Catalog No. Quantity
C155825G 25 g
1 options

Catalog No. C155825G

Supplier: TCI America C155825G

Only null left

Chemical Identifiers

CAS 608-23-1
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00060689
InChI Key NVLHGZIXTRYOKT-UHFFFAOYSA-N
Synonym 3-chloro-o-xylene, 2,3-dimethylchlorobenzene, benzene, chlorodimethyl, 3-chloro-1,2-dimethylbenzene, 3-chloro-ortho-xylene, benzene, 1-chloro-2,3-dimethyl, xylyl chloride, chloro o-xylene, mono-chlorxylol, chloro-ortho-xylene
PubChem CID 32884
IUPAC Name 1-chloro-2,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)Cl)C

Specifications

Color Yellow
Boiling Point 190°C
Quantity 25 g
Physical Form Clear Liquid at 20°C
Formula Weight 140.61
Percent Purity ≥96.0% (GC)
Chemical Name or Material 3-Chloro-o-xylene