missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-N,N-dimethylaniline 97.0+%, TCI America™

Catalog No. p-7125884
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C31925G 5 g
C319225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C31925G Supplier TCI America Supplier No. C31925G

Chemical Identifiers

CAS 6848-13-1
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00018456
InChI Key CHHCCYVOJBBCIY-UHFFFAOYSA-N
PubChem CID 23285
IUPAC Name 3-chloro-N,N-dimethylaniline
SMILES CN(C)C1=CC(=CC=C1)Cl

Specifications

Boiling Point 234°C
Quantity 5 g
Synonym n,n-dimethyl-3-chloroaniline, 3-chloro-nn-dimethylaniline, benzenamine, 3-chloro-n,n-dimethyl, m-chloro-n,n-dimethylaniline, benzenamine,3-chloro-n,n-dimethyl, 3-chlorophenyl dimethylamine, pubchem3637, 3-chloranyl-n,n-dimethyl-aniline, 3-n,n-dimethylamino phenyl chloride
Formula Weight 155.63
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Chloro-N,N-dimethylaniline