missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-N-Fmoc-L-phenylalanine, 95%

Catalog No. p-7056979
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6623703 1 g
AAH6623706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6623703 Supplier Thermo Scientific Chemicals Supplier No. H6623703
Only null left

CAS: 198560-44-0 | C24H20ClNO4 | 421.877 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 198560-44-0
Molecular Formula C24H20ClNO4
Molecular Weight (g/mol) 421.877
MDL Number MFCD00672548
InChI Key UOZAKKJRIKXQPY-QFIPXVFZSA-N
Synonym fmoc-l-3-chlorophenylalanine, fmoc-3-chloro-l-phenylalanine, fmoc-phe 3-cl-oh, fmoc-l-phe 3-cl-oh, l-3-3-chlorophenyl-n-fmoc-alanine, fmoc-l-3-chlorophe, l-3-chlorophenyl-n-fmoc-alanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-3-chlorophenyl propanoic acid, 2s-3-3-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-3-chlorophenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid
PubChem CID 2761466
IUPAC Name (2S)-3-(3-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC(=CC=C4)Cl)C(=O)O

Specifications

Quantity 1 g
Formula Weight 421.88
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Chloro-N-Fmoc-L-phenylalanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only