missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-DL-mandelic Acid 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 115°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Physical Form | Crystal-Powder at 20°C |
| Formula Weight | 186.59 |
| Percent Purity | ≥97.0% (GC,T) |
| Chemical Name or Material | 3-Chloro-DL-mandelic Acid |
Safety and Handling
| TSCA | No |