missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-alpha-toluenesulfonyl chloride, 96%

Catalog No. AAB2407303
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2407303 1 g
AAB2407306 5 g
AAB2407314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2407303 Supplier Thermo Scientific Chemicals Supplier No. B2407303

CAS: 24974-73-0 | C7H6Cl2O2S | 225.083 g/mol

3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloro-alpha-toluenesulfonyl chloride is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Store under dry inert gas. It is hygroscopic in nature. Incompatible with oxidizing agents. It is sensitive to moisture.

Chemical Identifiers

CAS 24974-73-0
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.083
MDL Number MFCD02683111
InChI Key LXTGNVLBPVVMSL-UHFFFAOYSA-N
Synonym 3-chlorophenyl methanesulfonyl chloride, 3-chloro-phenyl-methanesulfonyl chloride, chloro 3-chlorophenyl methyl sulfone, benzenemethanesulfonyl chloride, 3-chloro, pubchem5501, acmc-20an5u, 3-chlorobenzylsulfonyl chloride, 3-chlorophenyl methanesulfonylchloride, benzenemethanesulfonylchloride,3-chloro, benzenemethanesulfonylchloride, 3-chloro
PubChem CID 2757802
IUPAC Name (3-chlorophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC(=C1)Cl)CS(=O)(=O)Cl

Specifications

Melting Point 69°C to 75°C
Quantity 1 g
UN Number UN3261
Formula Weight 225.09
Percent Purity 96%
Chemical Name or Material 3-Chloro-alpha-toluenesulfonyl chloride
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only