missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-alpha-methylbenzyl Alcohol 98.0+%, TCI America™

Catalog No. C138210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C138210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C138210G Supplier TCI America Supplier No. C138210G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6939-95-3
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00021864
InChI Key QYUQVBHGBPRDKN-UHFFFAOYSA-N
Synonym 1-3-chlorophenyl ethanol, 3-chloro-alpha-methylbenzyl alcohol, 1-3-chlorophenyl ethan-1-ol, 1-3-chlorophenyl-1-ethanol, 3-chlorophenylethanol, acmc-1cap8, 1-m-chlorophenyl ethanol, 3'-chloro-1-phenylethanol, 1-3-chlorophenyl-ethanol, 1-3-chlorophenyl ethyl alcohol
PubChem CID 97779
IUPAC Name 1-(3-chlorophenyl)ethanol
SMILES CC(C1=CC(=CC=C1)Cl)O

Specifications

Color Yellow
Boiling Point 246°C
Quantity 10 g
Formula Weight 156.61
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Chloro-alpha-methylbenzyl Alcohol