missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-7-hydroxy-4-methylcoumarin 98.0+%, TCI America™
$227.41 - $823.90
Chemical Identifiers
| CAS | 6174-86-3 |
|---|---|
| Molecular Formula | C10H7ClO3 |
| Molecular Weight (g/mol) | 210.61 |
| MDL Number | MFCD00006851 |
| InChI Key | ODZHLDRQCZXQFQ-UHFFFAOYSA-N |
| Synonym | chlorferone, 3-chloro-7-hydroxy-4-methyl-2h-chromen-2-one, 3-chloro-4-methyl-7-hydroxycoumarin, chlorferron, 3-chloro-7-hydroxy-4-methylcoumarin, 7-hydroxy-4-methyl-3-chlorocoumarin, chlorferon, 3-chloro-4-methylumbelliferone, 3-chloro-7-hydroxy-4-methyl-2-benzopyrone, 2h-1-benzopyran-2-one, 3-chloro-7-hydroxy-4-methyl |
| PubChem CID | 5355079 |
| ChEBI | CHEBI:38620 |
| IUPAC Name | 3-chloro-7-hydroxy-4-methyl-2H-chromen-2-one |
| SMILES | CC1=C(Cl)C(=O)OC2=CC(O)=CC=C12 |
Chemical Identifiers
| 6174-86-3 | |
| 210.61 | |
| ODZHLDRQCZXQFQ-UHFFFAOYSA-N | |
| 5355079 | |
| 3-chloro-7-hydroxy-4-methyl-2H-chromen-2-one |
| C10H7ClO3 | |
| MFCD00006851 | |
| chlorferone, 3-chloro-7-hydroxy-4-methyl-2h-chromen-2-one, 3-chloro-4-methyl-7-hydroxycoumarin, chlorferron, 3-chloro-7-hydroxy-4-methylcoumarin, 7-hydroxy-4-methyl-3-chlorocoumarin, chlorferon, 3-chloro-4-methylumbelliferone, 3-chloro-7-hydroxy-4-methyl-2-benzopyrone, 2h-1-benzopyran-2-one, 3-chloro-7-hydroxy-4-methyl | |
| CHEBI:38620 | |
| CC1=C(Cl)C(=O)OC2=CC(O)=CC=C12 |
Specifications
| 6174-86-3 | |
| C10H7ClO3 | |
| 1 g | |
| ODZHLDRQCZXQFQ-UHFFFAOYSA-N | |
| 3-chloro-7-hydroxy-4-methyl-2H-chromen-2-one | |
| 5355079 | |
| 210.61 | |
| Crystalline Powder |
| 243°C | |
| MFCD00006851 | |
| chlorferone, 3-chloro-7-hydroxy-4-methyl-2h-chromen-2-one, 3-chloro-4-methyl-7-hydroxycoumarin, chlorferron, 3-chloro-7-hydroxy-4-methylcoumarin, 7-hydroxy-4-methyl-3-chlorocoumarin, chlorferon, 3-chloro-4-methylumbelliferone, 3-chloro-7-hydroxy-4-methyl-2-benzopyrone, 2h-1-benzopyran-2-one, 3-chloro-7-hydroxy-4-methyl | |
| CC1=C(Cl)C(=O)OC2=CC(O)=CC=C12 | |
| 210.61 | |
| CHEBI:38620 | |
| ≥98.0% (T) | |
| 3-Chloro-7-hydroxy-4-methylcoumarin |
Safety and Handling
TSCA : Yes