missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-6-phenylpyridazine 98.0+%, TCI America™
$221.83 - $684.11
Chemical Identifiers
| CAS | 20375-65-9 |
|---|---|
| Molecular Formula | C10H7ClN2 |
| Molecular Weight (g/mol) | 190.63 |
| MDL Number | MFCD00075253 |
| InChI Key | BUBRFWDEAVIFMV-UHFFFAOYSA-N |
| PubChem CID | 88515 |
| IUPAC Name | 3-chloro-6-phenylpyridazine |
| SMILES | C1=CC=C(C=C1)C2=NN=C(C=C2)Cl |
Chemical Identifiers
| 20375-65-9 | |
| 190.63 | |
| BUBRFWDEAVIFMV-UHFFFAOYSA-N | |
| 3-chloro-6-phenylpyridazine |
| C10H7ClN2 | |
| MFCD00075253 | |
| 88515 | |
| C1=CC=C(C=C1)C2=NN=C(C=C2)Cl |
Specifications
| 20375-65-9 | |
| C10H7ClN2 | |
| 1 g | |
| C1=CC=C(C=C1)C2=NN=C(C=C2)Cl | |
| 190.63 | |
| 190.63 | |
| Crystalline Powder |
| 159°C | |
| MFCD00075253 | |
| BUBRFWDEAVIFMV-UHFFFAOYSA-N | |
| 3-chloro-6-phenylpyridazine | |
| 88515 | |
| ≥98.0% (GC) | |
| 3-Chloro-6-phenylpyridazine |
Safety and Handling
TSCA : No