missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-6-phenylpyridazine 98.0+%, TCI America™

Catalog No. C34401G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34401G 1 g
C34405G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34401G Supplier TCI America Supplier No. C34401G
Only null left

Chemical Identifiers

CAS 20375-65-9
Molecular Formula C10H7ClN2
Molecular Weight (g/mol) 190.63
MDL Number MFCD00075253
InChI Key BUBRFWDEAVIFMV-UHFFFAOYSA-N
PubChem CID 88515
IUPAC Name 3-chloro-6-phenylpyridazine
SMILES C1=CC=C(C=C1)C2=NN=C(C=C2)Cl

Specifications

Melting Point 159°C
Quantity 1 g
Formula Weight 190.63
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Chloro-6-phenylpyridazine