missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-6-methoxypyridazine 97.0+%, TCI America™
$128.98 - $510.16
Chemical Identifiers
| CAS | 1722-10-7 |
|---|---|
| Molecular Formula | C5H5ClN2O |
| Molecular Weight (g/mol) | 144.56 |
| MDL Number | MFCD00006467 |
| InChI Key | XBJLKXOOHLLTPG-UHFFFAOYSA-N |
| PubChem CID | 74403 |
| IUPAC Name | 3-chloro-6-methoxypyridazine |
| SMILES | COC1=CC=C(Cl)N=N1 |
Chemical Identifiers
| 1722-10-7 | |
| 144.56 | |
| XBJLKXOOHLLTPG-UHFFFAOYSA-N | |
| 3-chloro-6-methoxypyridazine |
| C5H5ClN2O | |
| MFCD00006467 | |
| 74403 | |
| COC1=CC=C(Cl)N=N1 |
Specifications
| 1722-10-7 | |
| White-Yellow | |
| C5H5ClN2O | |
| 5 g | |
| XBJLKXOOHLLTPG-UHFFFAOYSA-N | |
| 3-chloro-6-methoxypyridazine | |
| 74403 | |
| ≥97.0% (GC) | |
| 3-Chloro-6-methoxypyridazine |
| 91°C | |
| 175°C | |
| MFCD00006467 | |
| pyridazine, 3-chloro-6-methoxy, 3-chloro-6-methoxy-pyridazine, 6-chloro-3-methoxypyridazine, pyridazine,3-chloro-6-methoxy, 6-chloropyridazin-3-yl methyl ether, 3-chloro-6-methoxypyridazine, zlchem 731, acmc-1bvtl, 3-methoxy-6-chloropyridazine, 3-chloro-6-methoxy pyridazine | |
| COC1=CC=C(Cl)N=N1 | |
| 144.56 | |
| 144.56 | |
| Crystalline Powder |
Safety and Handling
TSCA : No