missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-6-methoxypyridazine, 97%

CAS: 1722-10-7 | C5H5ClN2O | 144.56 g/mol

$111.58 - $1458.14

Chemical Identifiers

CAS 1722-10-7
Molecular Formula C5H5ClN2O
Molecular Weight (g/mol) 144.56
MDL Number MFCD00006467
InChI Key XBJLKXOOHLLTPG-UHFFFAOYSA-N
PubChem CID 74403
IUPAC Name 3-chloro-6-methoxypyridazine
SMILES COC1=CC=C(Cl)N=N1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1417306
View Documents
Thermo Scientific Chemicals
A1417306
5 g
Each for $111.58
Only null left
 
AAA1417314
View Documents
Thermo Scientific Chemicals
A1417314
25 g
Each for $354.82
Only null left
 
AAA1417322
View Documents
Thermo Scientific Chemicals
A1417322
100 g
Each for $1,458.14
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1722-10-7
144.56
XBJLKXOOHLLTPG-UHFFFAOYSA-N
3-chloro-6-methoxypyridazine
C5H5ClN2O
MFCD00006467
74403
COC1=CC=C(Cl)N=N1

Specifications

1722-10-7
C5H5ClN2O
5 g
pyridazine, 3-chloro-6-methoxy, 3-chloro-6-methoxy-pyridazine, 6-chloro-3-methoxypyridazine, pyridazine,3-chloro-6-methoxy, 6-chloropyridazin-3-yl methyl ether, 3-chloro-6-methoxypyridazine, zlchem 731, acmc-1bvtl, 3-methoxy-6-chloropyridazine, 3-chloro-6-methoxy pyridazine
COC1=CC=C(Cl)N=N1
144.56
144.56
3-Chloro-6-methoxypyridazine
90°C to 92°C
MFCD00006467
118854
XBJLKXOOHLLTPG-UHFFFAOYSA-N
3-chloro-6-methoxypyridazine
74403
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 217-019-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only